Ethyl-8-bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate ≥95%
Supplier: Matrix Scientific
046647-1G
046647-500MG
101876-398EA
375.38
USD
101876-398
101876-400
Ethyl-8-bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate ≥95%
Ethyl 8-bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Hazard: Irritant
Formula:
C₁₁H₈BrF₃N₂O₂ Melting Pt: 106…108 °C |
MDL Number:
MFCD12025881 CAS Number: 1038389-86-4 |
Specification Test Results
Melting point | 106-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...