2-Amino-5-bromoisonicotinic acid ≥98%

Supplier: Matrix Scientific

041229-1G
101850-992EA 38.46 USD
101850-992
2-Amino-5-bromoisonicotinic acid ≥98%
2-Amino-5-bromoisonicotinic acid

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.0200 g/mol
Melting Pt: 264…267 °C
MDL Number: MFCD09878357
CAS Number: 1000339-23-0

Order Now

Specification Test Results

Melting point 264-267°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR