8-Amino-4H-1,4-benzoxazin-3-one, Technical Grade

Supplier: Matrix Scientific

Synonyms: 8-Amino-2H-1,4-benzoxazin-3(4H)-one

041227-500MG
101850-986EA 200 USD
101850-986
8-Amino-4H-1,4-benzoxazin-3-one, Technical Grade
8-Amino-4H-1,4-benzoxazin-3-one

Hazard: Irritant

Formula: C₈H₈N₂O₂
MW: 164.17 g/mol
Melting Pt: 300 °C (dec.)
MDL Number: MFCD09998373
CAS Number: 321126-82-3

Order Now

Specification Test Results

Melting point 300°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR