4-(Dimethylamino)benzhydrazide

Supplier: Matrix Scientific

029691-500MG
101828-664EA 146.15 USD
101828-664
4-(Dimethylamino)benzhydrazide
4-(Dimethylamino)benzhydrazide

Hazard: Irritant

Formula: C₉H₁₃N₃O
Melting Pt: 174…175 °C
MDL Number: MFCD00014761
CAS Number: 19353-92-5

Order Now

Specification Test Results

Melting point 174-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR