4-Benzyloxybenzhydrazide

Supplier: Matrix Scientific

Synonyms: 4-(Benzyloxy)benzohydrazide

024022-500MG
101817-246EA 35.38 USD
101817-246
4-Benzyloxybenzhydrazide
4-Benzyloxybenzhydrazide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MW: 242.2732 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00564772
CAS Number: 128958-65-6

Order Now

Specification Test Results

Melting point 140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR