1,2,4,5-Tetrakis(trifluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

023798-500MG
101816-774EA 200 USD
101816-774
1,2,4,5-Tetrakis(trifluoromethyl)benzene ≥97%
1,2,4,5-Tetrakis(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₁₀H₂F₁₂
Melting Pt: 72…74 °C
MDL Number: MFCD08458697
CAS Number: 320-23-0

Order Now

Specification Test Results

Melting point 72-74°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR