2-Acetamidoisonicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Acetylaminoisonicotinic acid

023793-5G
101816-758EA 178.25 USD
101816-758
2-Acetamidoisonicotinic acid ≥97%
2-Acetamidoisonicotinic acid

Hazard: Irritant

Formula: C₈H₈N₂O₃
MW: 180.1600 g/mol
Melting Pt: 286…287 °C
MDL Number: MFCD00233713
CAS Number: 54221-95-3

Order Now

Specification Test Results

Melting point 286-287°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR