(2-Bromopyridin-3-yl)carbamic acid-tert-butyl ester ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (2-bromopyridin-3-yl)carbamate

023790-250MG
101816-752EA 112.31 USD
101816-752
(2-Bromopyridin-3-yl)carbamic acid-tert-butyl ester ≥97%
(2-Bromopyridin-3-yl)carbamic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O₂
MW: 273.14 g/mol
Melting Pt: 84…85 °C
MDL Number: MFCD07367911
CAS Number: 116026-98-3

Order Now

Specification Test Results

Melting point 84-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR