2-Acetylamino-4-methylpyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Acetamido-4-methylpyridineN-(4-Methyl-2-pyridyl)acetamideN-(4-Methylpyridin-2-yl)acetamide2-Acetamido-4-picoline

023783-250MG
101816-734EA 209.23 USD
101816-734
2-Acetylamino-4-methylpyridine ≥97%
2-Acetylamino-4-methylpyridine

Hazard: Irritant

Formula: C₈H₁₀N₂O
MW: 150.1778 g/mol
Melting Pt: 165…166 °C
MDL Number: MFCD00234432
CAS Number: 5327-32-2

Order Now

Specification Test Results

Melting point 165-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR