6-Methoxypyridine-2-carboxylic acid amide ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Methoxypicolinamide
023781-250MG
101816-730EA
273.85
USD
101816-730
6-Methoxypyridine-2-carboxylic acid amide ≥97%
6-Methoxypyridine-2-carboxylic acid amide
Hazard: Irritant
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 108…109 °C |
MDL Number:
MFCD08062921 CAS Number: 98276-69-8 |
Specification Test Results
Melting point | 108-109°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...