7-(Trifluoromethyl)quinoline ≥97%

Supplier: Matrix Scientific

023763-1G
101816-670EA 189.23 USD
101816-670
7-(Trifluoromethyl)quinoline ≥97%
7-(Trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₆F₃N
Melting Pt: 65…67 °C
MDL Number: MFCD00833760
CAS Number: 325-14-4

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR