(3-Bromo-4,5-dihydroisoxazol-5-ylmethyl)carbamic acid tert-butyl ester ≥97%

Supplier: Matrix Scientific

023762-1G
101816-668EA 138.46 USD
101816-668
(3-Bromo-4,5-dihydroisoxazol-5-ylmethyl)carbamic acid tert-butyl ester ≥97%
(3-Bromo-4,5-dihydroisoxazol-5-ylmethyl)carbamic acid tert-butyl ester

Hazard: Irritant

Formula: C₉H₁₅BrN₂O₃
MW: 279.14 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD08741480
CAS Number: 109770-82-3

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR