4-Methoxypyridine-2-carboxylic acid hydrazide ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Methoxypicolinohydrazide

023724-250MG
101816-574EA 229.23 USD
101816-574
4-Methoxypyridine-2-carboxylic acid hydrazide ≥98%
4-Methoxypyridine-2-carboxylic acid hydrazide

Hazard: Irritant

Formula: C₇H₉N₃O₂
MW: 167.17 g/mol
Melting Pt: 157…159 °C
MDL Number: MFCD07779513
CAS Number: 187973-18-8

Order Now

Specification Test Results

Melting point 157-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR