6-Iodopyridin-2-ylamine ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Iodopyridin-2-amine

023722-250MG
101816-570EA 192.31 USD
101816-570
6-Iodopyridin-2-ylamine ≥98%
6-Iodopyridin-2-ylamine

Hazard: Irritant

Formula: C₅H₅IN₂
MW: 220.01 g/mol
Melting Pt: 103…104 °C
MDL Number: MFCD07779511
CAS Number: 88511-25-5

Order Now

Specification Test Results

Melting point 103-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR