3-Chloro-2-(2-ethoxyethoxy)aniline

Supplier: Matrix Scientific

021872-500MG
101812-906EA 315.38 USD
101812-906
3-Chloro-2-(2-ethoxyethoxy)aniline
3-Chloro-2-(2-ethoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₂
Melting Pt: 69…70 °C
MDL Number: MFCD08687290
CAS Number: 946772-33-4

Order Now

Specification Test Results

Melting point 69-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR