5-Bromo-6-chloronicotinic acid ≥98%
Supplier: Matrix Scientific
021370-5G
021370-1G
021370-25G
101811-874EA
16.92
USD
101811-874
102832-910
101811-876
5-Bromo-6-chloronicotinic acid ≥98%
5-Bromo-6-chloronicotinic acid
Hazard: Irritant
Formula:
C₆H₃BrClNO₂ MW: 236.45 g/mol Melting Pt: 166…168 °C |
MDL Number:
MFCD01927098 CAS Number: 29241-62-1 |
Specification Test Results
Melting point | 166-168°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...