(2-Bromo-4-pyridinyl)methanol ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Bromopyridine-4-methanol
, 2-Bromo-4-pyridinemethanol
021365-1G
021365-25G
021365-5G
102551-018EA
287.69
USD
102551-018
101811-858
101811-856
(2-Bromo-4-pyridinyl)methanol ≥97%
(2-Bromo-4-pyridinyl)methanol
Hazard: Irritant
Formula:
C₆H₆BrNO MW: 188.02 g/mol Melting Pt: 58…61 °C |
MDL Number:
MFCD04039313 CAS Number: 118289-16-0 |
Specification Test Results
Melting point | 58-61°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...