1,6-Naphthyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
1,6-Diazanaphthalene
, Pyrido[4,3-b]pyridine
021364-250MG
101811-854EA
113.85
USD
101811-854
1,6-Naphthyridine ≥98%
1,6-Naphthyridine
Hazard: Irritant
Formula:
C₈H₆N₂ MW: 130.15 g/mol Melting Pt: 231…233 °C |
MDL Number:
MFCD00059750 CAS Number: 253-72-5 |
Specification Test Results
Melting point | 231-233°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...