2-Chloro-4-methoxybenzyl bromide ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2-chloro-4-methoxybenzene

021048-1G 021048-5G
101811-268EA 63.08 USD
101811-268 101811-270
2-Chloro-4-methoxybenzyl bromide ≥95%
2-Chloro-4-methoxybenzyl bromide

Hazard: Toxic/Irritant/Refrigerate

Formula: C₈H₈BrClO
MW: 235.5055 g/mol
Melting Pt: 57…58 °C
MDL Number: MFCD08445696
CAS Number: 54788-17-9

Order Now

Specification Test Results

Melting point 57-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR