2-Chloro-5-fluorophenylacetonitrile ≥99%
Supplier: Matrix Scientific
011283-1G
011283-5G
101800-330EA
192.31
USD
101800-330
101800-328
2-Chloro-5-fluorophenylacetonitrile ≥99%
2-Chloro-5-fluorophenylacetonitrile
Purity: 99%
CAS: 395675-23-7
MDL Number: MFCD06246270
Molecular Formula: C8H5ClFN
Molecular Weight: 169,59
Hazard: Irritant/Irritant-Harmful
Formula:
C₈H₅ClFN Melting Pt: 58…59 °C |
MDL Number:
MFCD06246270 CAS Number: 395675-23-7 |
Specification Test Results
Melting point | 58-59°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...