N-2-Fluorophenyl-3-(trifluoromethyl)-benzamide ≥98%
Supplier: Matrix Scientific
004944-5G
004944-1G
101788-080EA
52.31
USD
101788-080
101788-082
N-2-Fluorophenyl-3-(trifluoromethyl)-benzamide ≥98%
N-2-Fluorophenyl-3-(trifluoromethyl)-benzamide
Purity: 98%
CAS: 199461-55-7
MDL Number: MFCD00045085
Molecular Formula: C14H9F4NO
Molecular Weight: 283,23
Hazard: Irritant
Formula:
C₁₄H₉F₄NO Melting Pt: 105…107 °C |
MDL Number:
MFCD00045085 CAS Number: 199461-55-7 |
Specification Test Results
Melting point | 105-107°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...