1-(2-Cyanoethyl)-5-(trifluoromethyl)pyrid-2-(1H)-one
Supplier: Matrix Scientific
004871-1G
101787-934EA
49.23
USD
101787-934
1-(2-Cyanoethyl)-5-(trifluoromethyl)pyrid-2-(1H)-one
1-(2-Cyanoethyl)-5-(trifluoromethyl)pyrid-2-(1H)-one
CAS: 175277-71-1
MDL Number: MFCD00204218
Molecular Formula: C9H7F3N2O
Molecular Weight: 216,16
Hazard: Irritant
Formula:
C₉H₇F₃N₂O MW: 216.16 g/mol Melting Pt: 89…90 °C |
MDL Number:
MFCD00204218 CAS Number: 175277-71-1 |
Specification Test Results
Melting point | 89-90°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...