1-(2-Cyanoethyl)-3-(trifluoromethyl)pyrid-2-(1H)-one
Supplier: Matrix Scientific
004870-1G
101787-932EA
66.15
USD
101787-932
1-(2-Cyanoethyl)-3-(trifluoromethyl)pyrid-2-(1H)-one
1-(2-Cyanoethyl)-3-(trifluoromethyl)pyrid-2-(1H)-one
CAS: 175277-60-8
MDL Number: MFCD00204216
Molecular Formula: C9H7F3N2O
Molecular Weight: 216,16
Hazard: Irritant
Formula:
C₉H₇F₃N₂O MW: 216.16 g/mol Melting Pt: 88…89 °C |
MDL Number:
MFCD00204216 CAS Number: 175277-60-8 |
Specification Test Results
Melting point | 88-89°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...