3-Amino-4-(4-morpholino)benzotrifluoride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(4-Morpholinyl)-5-(trifluoromethyl)aniline2-Morpholino-5-(trifluoromethyl)aniline

004799-1G
101787-834EA 51.75 USD
101787-834
3-Amino-4-(4-morpholino)benzotrifluoride ≥97%
3-Amino-4-(4-morpholino)benzotrifluoride

Hazard: Toxic

Formula: C₁₁H₁₃F₃N₂O
MW: 246.23 g/mol
Melting Pt: 127…128 °C
MDL Number: MFCD00042160
CAS Number: 784-57-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 127-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR