N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
Supplier: Matrix Scientific
004793-5G
004793-1G
101787-822EA
24.62
USD
101787-822
101787-824
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
Hazard: Irritant
Formula:
C₁₁H₁₂F₃N₃O₂ Melting Pt: 68…69 °C |
MDL Number:
MFCD00052613 CAS Number: 58315-38-1 |
Specification Test Results
Melting point | 68-69°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...