2-Fluoro-6-(1-pyrrolyl)benzonitrile
Supplier: Matrix Scientific
004789-1G
101787-812EA
209.23
USD
101787-812
2-Fluoro-6-(1-pyrrolyl)benzonitrile
2-Fluoro-6-(1-pyrrolyl)benzonitrile
Hazard: Irritant
Formula:
C₁₁H₇FN₂ Melting Pt: 67…69 °C |
MDL Number:
MFCD00052222 CAS Number: 148901-51-3 |
Specification Test Results
Melting point | 67-69°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...