4-Iodophenyl ether ≥98%
Supplier: Matrix Scientific
004704-5G
004704-25G
101787-704EA
253.85
USD
101787-704
101787-702
4-Iodophenyl ether ≥98%
4-Iodophenyl ether
Purity: 98%
CAS: 28896-49-3
MDL Number: MFCD00060667
Molecular Formula: C12H8I2O
Molecular Weight: 422,01
Hazard: Irritant
Formula:
C₁₂H₈I₂O Melting Pt: 144…146 °C |
MDL Number:
MFCD00060667 CAS Number: 28896-49-3 |
Specification Test Results
Melting point | 144-146°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...