2,4-Diamino-6-(4-Fluorophenyl)Pyrimidine

Supplier: Matrix Scientific

004673-1G
101787-666EA 67.69 USD
101787-666
2,4-Diamino-6-(4-Fluorophenyl)Pyrimidine
2,4-Diamino-6-(4-Fluorophenyl)Pyrimidine
Formula: C₁₀H₉FN₄
Melting Pt: 164…166 °C
MDL Number: MFCD00052076
CAS Number: 175137-25-4

Order Now

Specification Test Results

Melting point 164-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR