Thiophene-3-carboxylic acid hydrazide

Supplier: Matrix Scientific

Synonyms: Thiophene-3-carbohydrazide

004627-1G
101787-600EA 210.77 USD
101787-600
Thiophene-3-carboxylic acid hydrazide
Thiophene-3-carboxylic acid hydrazide

CAS: 39001-23-5
MDL Number: MFCD01310840
Molecular Formula: C5H6N2OS
Molecular Weight: 142,18
Hazard: Irritant

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Melting Pt: 126…128 °C
MDL Number: MFCD01310840
CAS Number: 39001-23-5

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR