2-(Methylamino)-5-(trifluoromethyl)-1,3,4-thiadiazole ≥97%

Supplier: Matrix Scientific

004518-1G
101787-378EA 100 USD
101787-378
2-(Methylamino)-5-(trifluoromethyl)-1,3,4-thiadiazole ≥97%
2-(Methylamino)-5-(trifluoromethyl)-1,3,4-thiadiazole

Hazard: Irritant

Formula: C₄H₄F₃N₃S
Melting Pt: 115…117 °C
MDL Number: MFCD00042321
CAS Number: 25366-22-7

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR