Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
004516-1G
101787-372EA
36.92
USD
101787-372
Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate ≥97%
Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
Purity: 97%
CAS: 149771-09-5
MDL Number: MFCD00052070
Molecular Formula: C8H8F3N3O2
Molecular Weight: 235,17
Hazard: Irritant
Formula:
C₈H₈F₃N₃O₂ MW: 235.17 g/mol Melting Pt: 179…180 °C |
MDL Number:
MFCD00052070 CAS Number: 149771-09-5 |
Specification Test Results
Melting point | 179-180°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...