2-Iodo-4-methyl-6-nitrophenol

Supplier: Matrix Scientific

004483-5G
101787-310EA 153.85 USD
101787-310
2-Iodo-4-methyl-6-nitrophenol
2-Iodo-4-methyl-6-nitrophenol

Hazard: Irritant

Formula: C₇H₆INO₃
Melting Pt: 82…84 °C
MDL Number: MFCD00060537
CAS Number: 69492-91-7

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR