3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole ≥95%
Supplier: Matrix Scientific
004478-250MG
004478-1G
101787-302EA
73.85
USD
101787-302
101787-300
3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole ≥95%
3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole
Hazard: Irritant
Formula:
C₁₀H₆ClF₃N₂O Melting Pt: 25…27 °C |
MDL Number:
MFCD00084969 CAS Number: 175205-63-7 |
Specification Test Results
Melting point | 25-27°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...