3-Iodo-4-methoxybiphenyl ≥98%
Supplier: Matrix Scientific
004463-5G
004463-1G
101787-274EA
315.38
USD
101787-274
101787-272
3-Iodo-4-methoxybiphenyl ≥98%
3-Iodo-4-methoxybiphenyl
Purity: 98%
CAS: 91718-20-6
MDL Number: MFCD00079763
Molecular Formula: C13H11IO
Molecular Weight: 310,14
Hazard: Irritant
Formula:
C₁₃H₁₁IO Melting Pt: 89…91 °C |
MDL Number:
MFCD00079763 CAS Number: 91718-20-6 |
Specification Test Results
Melting point | 89-91°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...