3-(Pentafluorophenyl)propionic acid

Supplier: Matrix Scientific

Synonyms: 3-(Perfluorophenyl)propanoic acid

004421-1G 004421-5G
101787-214EA 304.62 USD
101787-214 101787-212
3-(Pentafluorophenyl)propionic acid
3-(Pentafluorophenyl)propionic acid

CAS: 2002-92-8
MDL Number: MFCD00020516
Molecular Formula: C9H5F5O2
Molecular Weight: 240,13
Hazard: Corrosive

Formula: C₉H₅F₅O₂
MW: 240.13 g/mol
Melting Pt: 91…92 °C
MDL Number: MFCD00020516
CAS Number: 2002-92-8

Order Now

Specification Test Results

Melting point 91-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR