3-(Heptafluoro-n-propyl)-5-methyl-4-nitropyrazole
Supplier: Matrix Scientific
004371-5G
101787-114EA
144.62
USD
101787-114
3-(Heptafluoro-n-propyl)-5-methyl-4-nitropyrazole
3-(Heptafluoro-n-propyl)-5-methyl-4-nitropyrazole
CAS: 82633-69-0
MDL Number: MFCD00155841
Molecular Formula: C7H4F7N3O2
Molecular Weight: 295,12
Hazard: Irritant
Formula:
C₇H₄F₇N₃O₂ Melting Pt: 97…98 °C |
MDL Number:
MFCD00155841 CAS Number: 82633-69-0 |
Specification Test Results
Melting point | 97-98°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...