MATR004311

004311-100G
101787-086EA 49.69 USD
101787-086
MATR004311
Purity: 98%
CAS: 3144-09-0
MDL Number: MFCD00007940
Molecular Formula: CH5NO2S
Molecular Weight: 95,12
TCA listed: T
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR