3,4-Methylenedioxybenzhydrazide ≥98%
Supplier: Matrix Scientific
Synonyms:
Benzo[d][1,3]dioxole-5-carbohydrazide
004297-1G
004297-5G
101787-058EA
213.85
USD
101787-058
101787-056
3,4-Methylenedioxybenzhydrazide ≥98%
3,4-Methylenedioxybenzhydrazide
Purity: 98%
CAS: 22026-39-7
MDL Number: MFCD00060505
Molecular Formula: C8H8N2O3
Molecular Weight: 180,16
Hazard: Irritant
Formula:
C₈H₈N₂O₃ MW: 180.16 g/mol Melting Pt: 174 °C |
MDL Number:
MFCD00060505 CAS Number: 22026-39-7 |
Specification Test Results
Melting point | 174°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...