4-(Diethylamino)benzhydrazide

Supplier: Matrix Scientific

004273-5G 004273-1G
101787-036EA 186.15 USD
101787-036 101787-034
4-(Diethylamino)benzhydrazide
4-(Diethylamino)benzhydrazide

Hazard: Irritant

Formula: C₁₁H₁₇N₃O
Melting Pt: 99…101 °C
MDL Number: MFCD01310809
CAS Number: 100139-54-6

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR