4-[4-(Methylthio)phenoxy]-3-nitrobenzotrifluoride
Supplier: Matrix Scientific
004099-1G
004099-250MG
101786-828EA
47.69
USD
101786-828
101786-830
4-[4-(Methylthio)phenoxy]-3-nitrobenzotrifluoride
4-[4-(Methylthio)phenoxy]-3-nitrobenzotrifluoride
CAS: 56191-23-2
MDL Number: MFCD00052261
Molecular Formula: C14H10F3NO3S
Molecular Weight: 329,3
Hazard: Irritant
Formula:
C₁₄H₁₀F₃NO₃S Melting Pt: 41…43 °C |
MDL Number:
MFCD00052261 CAS Number: 56191-23-2 |
Specification Test Results
Melting point | 41-43°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...