3,5-Dimethoxy-4-hydroxyphenylacetonitrile ≥98%
Supplier: Matrix Scientific
003991-5G
003991-1G
101786-598EA
126.15
USD
101786-598
101786-600
3,5-Dimethoxy-4-hydroxyphenylacetonitrile ≥98%
3,5-Dimethoxy-4-hydroxyphenylacetonitrile
Purity: 98%
CAS: 42973-55-7
MDL Number: MFCD00060308
Molecular Formula: C10H11NO3
Molecular Weight: 193,2
Hazard: Irritant/Irritant-Harmful
Formula:
C₁₀H₁₁NO₃ Melting Pt: 68…70 °C |
MDL Number:
MFCD00060308 CAS Number: 42973-55-7 |
Specification Test Results
Melting point | 68-70°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...