4-Fluoro-N-[4-(trifluoromethyl)phenyl]benzamide ≥97%
Supplier: Matrix Scientific
003747-5G
003747-1G
101786-156EA
72.31
USD
101786-156
101786-158
4-Fluoro-N-[4-(trifluoromethyl)phenyl]benzamide ≥97%
4-Fluoro-N-[4-(trifluoromethyl)phenyl]benzamide
Hazard: Irritant
Formula:
C₁₄H₉F₄NO Melting Pt: 153…154 °C |
MDL Number:
MFCD00045094 CAS Number: 2054-01-5 |
Specification Test Results
Melting point | 153-154°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...