2-Furoyltrifluoroacetone ≥99%

Supplier: Matrix Scientific

Synonyms: 4,4,4-Trifluoro-1-(furan-2-yl)butane-1,3-dione4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione

003548-5G 003548-25G
101785-852EA 267.69 USD
101785-852 101785-850
2-Furoyltrifluoroacetone ≥99%
2-Furoyltrifluoroacetone

Hazard: Irritant

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Melting Pt: 20…21 °C
MDL Number: MFCD00020935
CAS Number: 326-90-9

Order Now

Specification Test Results

Melting point 20-21°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR