4,4'-Diacetylbiphenyl ≥98%

Supplier: Matrix Scientific

003518-25G 003518-5G
101785-834EA 39.1 USD
101785-834 101785-832
4,4'-Diacetylbiphenyl ≥98%
4,4'-Diacetylbiphenyl

Hazard: Irritant

Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
Melting Pt: 194 °C
MDL Number: MFCD00017248
CAS Number: 787-69-9

Order Now

Specification Test Results

Melting point 194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR