(4-Bromotetrafluorophenyl)hydrazine ≥97%
Supplier: Matrix Scientific
003470-5G
003470-100G
003470-25G
101785-748EA
46.15
USD
101785-748
101785-750
101785-752
(4-Bromotetrafluorophenyl)hydrazine ≥97%
(4-Bromotetrafluorophenyl)hydrazine
Purity: 97%
CAS: 2797-79-7
MDL Number: MFCD01075256
Molecular Formula: C6H3BrF4N2
Molecular Weight: 259,01
Hazard: Toxic
Formula:
C₆H₃BrF₄N₂ Melting Pt: 79…80 °C |
MDL Number:
MFCD01075256 CAS Number: 2797-79-7 |
Specification Test Results
Melting point | 79-80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...