Ethyl-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-methyl-2-(4-(trifluoromethyl)phenyl)thiazole-5-carboxylate
003391-25G
003391-5G
101785-658EA
47.69
USD
101785-658
101785-656
Ethyl-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Ethyl-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
CAS: 175277-03-9
MDL Number: MFCD00068104
Molecular Formula: C14H12F3NO2S
Molecular Weight: 315,32
Hazard: Irritant
Formula:
C₁₄H₁₂F₃NO₂S MW: 315.32 g/mol Melting Pt: 85…86 °C |
MDL Number:
MFCD00068104 CAS Number: 175277-03-9 |
Specification Test Results
Melting point | 85-86°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...