Diphenylmethane-4-carboxylic acid ≥98%
Supplier: Matrix Scientific
003370-1G
003370-5G
101785-632EA
278.46
USD
101785-632
101785-630
Diphenylmethane-4-carboxylic acid ≥98%
Diphenylmethane-4-carboxylic acid
Purity: 98%
CAS: 620-86-0
MDL Number: MFCD00016540
Molecular Formula: C14H12O2
Molecular Weight: 212,25
Hazard: Irritant
Formula:
C₁₄H₁₂O₂ Melting Pt: 162…164 °C |
MDL Number:
MFCD00016540 CAS Number: 620-86-0 |
Specification Test Results
Melting point | 162-164°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...