MATR003270

003270-25G 003270-100G
101785-486EA 73.23 USD
101785-486 101785-484
MATR003270
Purity: 98%
CAS: 133745-75-2
MDL Number: MFCD00144885
Molecular Formula: C12H10FNO4S2
Molecular Weight: 315,34
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR