Bis(4-cyanophenyl)ether

Supplier: Matrix Scientific

Synonyms: 4,4'-Oxydibenzonitrile4,4'-Dicyanodiphenyl Ether

003205-1G
101785-406EA 93.85 USD
101785-406
Bis(4-cyanophenyl)ether
Bis(4-cyanophenyl)ether

CAS: 6508-04-9
MDL Number: MFCD00070640
Molecular Formula: C14H8N2O
Molecular Weight: 220,23
Boiling Point: 220°C/1 Torr
TCA listed: T
Hazard: Irritant/Irritant-Harmful

Formula: C₁₄H₈N₂O
MW: 220.23 g/mol
Boiling Pt: 220 °C (1 Torr)
Melting Pt: 176…178 °C°C
CAS Number: 6508-04-9

Order Now

Specification Test Results

Melting point 184-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR