3-(2,6-Dichlorophenyl)propionic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
3-(2,6-Dichlorophenyl)propanoic acid
003097-5G
003097-25G
101785-232EA
160.82
USD
101785-232
101785-230
3-(2,6-Dichlorophenyl)propionic acid ≥98%
3-(2,6-Dichlorophenyl)propionic acid
Purity: 98%
CAS: 51656-68-9
MDL Number: MFCD01310807
Molecular Formula: C9H8Cl2O2
Molecular Weight: 219,07
Hazard: Irritant
Formula:
C₉H₈Cl₂O₂ MW: 219.07 g/mol Melting Pt: 95…97 °C |
MDL Number:
MFCD01310807 CAS Number: 51656-68-9 |
Specification Test Results
Melting point | 95-97°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...